3-Cyanobenzaldehyde 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD19666-10GEA 11 EUR
BLDPBD19666-10G BLDPBD19666-5G BLDPBD19666-100G BLDPBD19666-25G
3-Cyanobenzaldehyde 98%
3-Cyanobenzaldehyde

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₈H₅NO
MW: 131,13 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00003344
CAS-nummer: 24964-64-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR