5-(Hydroxymethyl)pyrimidine 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD19689-250MGEA 19 EUR
BLDPBD19689-250MG BLDPBD19689-5G BLDPBD19689-10G BLDPBD19689-1G BLDPBD19689-25G
5-(Hydroxymethyl)pyrimidine 98%
5-(Hydroxymethyl)pyrimidine

Storage condition: Inert atmosphere, 2-8°C

Formule: C₅H₆N₂O
MW: 110,1100 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD03789623
CAS-nummer: 25193-95-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR