5-Cyano-2-methylpyridine 98%

BLDPBD20214-10GEA 46 EUR
BLDPBD20214-10G BLDPBD20214-100G BLDPBD20214-25G BLDPBD20214-5G BLDPBD20214-250MG BLDPBD20214-1G
5-Cyano-2-methylpyridine 98%
5-Cyano-2-methylpyridine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₇H₆N₂
MW: 118,14 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00038042
CAS-nummer: 3222-48-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR