4-Fluorobiphenyl 98%

BLDPBD20225-25GEA 157 EUR
BLDPBD20225-25G BLDPBD20225-5G BLDPBD20225-1G
4-Fluorobiphenyl 98%
4-Fluorobiphenyl

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₂H₉F
MW: 172,2 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00011650
CAS-nummer: 324-74-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR