(S)-1-Boc-2-ethylpiperazine 97%
|
|
BLDPBD20229-25GEA
1112
EUR
BLDPBD20229-25G
BLDPBD20229-10G
BLDPBD20229-100MG
BLDPBD20229-5G
BLDPBD20229-250MG
BLDPBD20229-1G
(S)-1-Boc-2-ethylpiperazine 97%
(S)-1-Boc-2-ethylpiperazine
Storage condition: Keep in dark place, Sealed in dry, 2-8°C
Formule:
C₁₁H₂₂N₂O₂ MW: 214,30 g/mol Opslagtemperatuur: Koelkast |
MDL-nummer:
MFCD07772100 CAS-nummer: 325145-35-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...