Boc-b-Alanine 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD20260-25GEA 13 EUR
BLDPBD20260-25G BLDPBD20260-1000G BLDPBD20260-500G BLDPBD20260-100G BLDPBD20260-5G BLDPBD20260-10G
Boc-b-Alanine 97%
Boc-b-Alanine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₈H₁₅NO₄
MW: 189,2 g/mol
Opslagtemperatuur: Kamertemperatuur
CAS-nummer: 3303-84-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR