4-(Trifluoromethoxy)phenyl isocyanate 98%

BLDPBD20373-5GEA 10 EUR
BLDPBD20373-5G BLDPBD20373-25G BLDPBD20373-1G
4-(Trifluoromethoxy)phenyl isocyanate 98%
4-(Trifluoromethoxy)phenyl isocyanate

Storage condition: Inert atmosphere, 2-8°C

Formule: C₈H₄F₃NO₂
MW: 203,12 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00035702
CAS-nummer: 35037-73-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR