Azetidine hydrochloride 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD20465-100GEA 445 EUR
BLDPBD20465-100G BLDPBD20465-25G BLDPBD20465-5G BLDPBD20465-10G BLDPBD20465-1G
Azetidine hydrochloride 95%
Azetidine hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formule: C₃H₈ClN
MW: 93,56 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00191762
CAS-nummer: 36520-39-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR