5-Acetylsalicylamide 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD20663-25GEA 18 EUR
BLDPBD20663-25G BLDPBD20663-10G BLDPBD20663-100G BLDPBD20663-500G
5-Acetylsalicylamide 98%
5-Acetylsalicylamide

Storage condition: Sealed in dry, Room Temperature

Formule: C₉H₉NO₃
MW: 179,18 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00049222
CAS-nummer: 40187-51-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR