Methyl-2-piperidinecarboxylate 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD20761-100GEA 61 EUR
BLDPBD20761-100G BLDPBD20761-10G
Methyl-2-piperidinecarboxylate 98%
Methyl-2-piperidinecarboxylate

Storage condition: Keep in dark place, Sealed in dry, Store in freezer, under -20°C

Formule: C₇H₁₃NO₂
MW: 143,1836 g/mol
Opslagtemperatuur: Vriezer
MDL-nummer: MFCD07364347
CAS-nummer: 41994-45-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR