1H-Benzimidazole-5-boronic acid pinacol ester 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD208143-1GEA 68 EUR
BLDPBD208143-1G BLDPBD208143-25G BLDPBD208143-5G BLDPBD208143-250MG BLDPBD208143-100MG
1H-Benzimidazole-5-boronic acid pinacol ester 95%
1H-Benzimidazole-5-boronic acid pinacol ester

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formule: C₁₃H₁₇BN₂O₂
MW: 244,10 g/mol
Opslagtemperatuur: Vriezer
MDL-nummer: MFCD11054041
CAS-nummer: 1007206-54-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR