1-Methylazetidine-3-carboxylic acid 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD208182-250MGEA 15 EUR
BLDPBD208182-250MG BLDPBD208182-1G BLDPBD208182-100MG BLDPBD208182-5G BLDPBD208182-25G
1-Methylazetidine-3-carboxylic acid 97%
1-Methylazetidine-3-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formule: C₅H₉NO₂
MW: 115,1300 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD11520621
CAS-nummer: 875629-26-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR