4-Chloro-5-methoxypyrimidine 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD208586-250MGEA 53 EUR
BLDPBD208586-250MG BLDPBD208586-100MG BLDPBD208586-1G BLDPBD208586-5G
4-Chloro-5-methoxypyrimidine 98%
4-Chloro-5-methoxypyrimidine

Storage condition: Inert atmosphere, 2-8°C

Formule: C₅H₅ClN₂O
MW: 144,5600 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD09966063
CAS-nummer: 695-85-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR