3-(Chloromethyl)-5-methylpyridine hydrochloride 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD208885-1GEA 14 EUR
BLDPBD208885-1G BLDPBD208885-0.025 BLDPBD208885-25G BLDPBD208885-10G BLDPBD208885-5G BLDPBD208885-250MG
3-(Chloromethyl)-5-methylpyridine hydrochloride 97%
3-(Chloromethyl)-5-methylpyridine hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formule: C7H9Cl2N
MW: 178,0600 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD11656303
CAS-nummer: 1007089-84-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR