1-(1-Boc-piperidino)-1H-pyrazole-4-boronic acid pinacol ester 98%

BLDPBD209079-100MGEA 6 EUR
BLDPBD209079-100MG BLDPBD209079-1G BLDPBD209079-25G BLDPBD209079-5G BLDPBD209079-0.025 BLDPBD209079-250MG BLDPBD209079-100G BLDPBD209079-10G
1-(1-Boc-piperidino)-1H-pyrazole-4-boronic acid pinacol ester 98%
1-(1-Boc-piperidino)-1H-pyrazole-4-boronic acid pinacol ester

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formule: C₁₉H₃₂BN₃O₄
MW: 377,2861 g/mol
Opslagtemperatuur: Vriezer
MDL-nummer: MFCD11112131
CAS-nummer: 877399-74-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR