1,8-Naftyleendiamine 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD20972-500GEA 120 EUR
BLDPBD20972-500G BLDPBD20972-100G BLDPBD20972-10G BLDPBD20972-25G
1,8-Naftyleendiamine 97%
1,8-Naftyleendiamine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₁₀H₁₀N₂
MW: 158,2 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00004033
CAS-nummer: 479-27-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR