3-Nitro-4-quinolinol 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD21052-100GEA 421 EUR
BLDPBD21052-100G BLDPBD21052-1G BLDPBD21052-5G BLDPBD21052-500G BLDPBD21052-25G BLDPBD21052-10G
3-Nitro-4-quinolinol 97%
3-Nitro-4-quinolinol

Storage condition: Inert atmosphere, Room Temperature

Formule: C₉H₆N₂O₃
MW: 190,1600 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD01820324
CAS-nummer: 50332-66-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR