1-(Tetrahydro-2H-pyran-4-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%

BLDPBD210595-25GEA 388 EUR
BLDPBD210595-25G BLDPBD210595-5G BLDPBD210595-1G BLDPBD210595-250MG
1-(Tetrahydro-2H-pyran-4-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%
1-(Tetrahydro-2H-pyran-4-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formule: C₁₄H₂₃BN₂O₃
MW: 278,1550 g/mol
Opslagtemperatuur: Vriezer
MDL-nummer: MFCD12033229
CAS-nummer: 1040377-03-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR