2-Aminomalonamide 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD211173-100GEA 39 EUR
BLDPBD211173-100G BLDPBD211173-250MG BLDPBD211173-1G BLDPBD211173-5G BLDPBD211173-25G BLDPBD211173-10G
2-Aminomalonamide 97%
2-Aminomalonamide

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₃H₇N₃O₂
MW: 117,1100 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00015932
CAS-nummer: 62009-47-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR