Ethyl-5-methyl-1H-imidazole-4-carboxylate 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD21156-500GEA 874 EUR
BLDPBD21156-500G BLDPBD21156-5G BLDPBD21156-25G BLDPBD21156-100G BLDPBD21156-10G BLDPBD21156-1G
Ethyl-5-methyl-1H-imidazole-4-carboxylate 98%
Ethyl-5-methyl-1H-imidazole-4-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formule: C₇H₁₀N₂O₂
MW: 154,1665 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00005199
CAS-nummer: 51605-32-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR