2-Iodo-1,3-dimethoxybenzene 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD212709-5GEA 46 EUR
BLDPBD212709-5G BLDPBD212709-1G BLDPBD212709-25G BLDPBD212709-250MG BLDPBD212709-10G BLDPBD212709-100G
2-Iodo-1,3-dimethoxybenzene 97%
2-Iodo-1,3-dimethoxybenzene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₈H₉IO₂
MW: 264,0603 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD05865214
CAS-nummer: 16932-44-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR