6,7-Dihydro-5H-quinoline-8-one 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD21524-5GEA 51 EUR
BLDPBD21524-5G BLDPBD21524-0.025G BLDPBD21524-10G BLDPBD21524-250MG BLDPBD21524-100G BLDPBD21524-1G BLDPBD21524-25G
6,7-Dihydro-5H-quinoline-8-one 98%
6,7-Dihydro-5H-quinoline-8-one

Storage condition: Sealed in dry, Room Temperature

Formule: C₉H₉NO
MW: 147,1700 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD03839916
CAS-nummer: 56826-69-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR