DL-2,4-Diaminobutyric acid dihydrochloride 97%
|
|
BLDPBD2158-250MGEA
11
EUR
BLDPBD2158-250MG
BLDPBD2158-5G
BLDPBD2158-10G
BLDPBD2158-100MG
BLDPBD2158-1G
DL-2,4-Diaminobutyric acid dihydrochloride 97%
DL-2,4-Diaminobutyric acid dihydrochloride
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formule:
C₄H₁₀N₂O₂·₂HCl MW: 191,1 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00012910 CAS-nummer: 65427-54-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...