tert-Butyl (1-(hydroxymethyl)cyclobutyl)carbamate 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD216361-5GEA 616 EUR
BLDPBD216361-5G BLDPBD216361-1G BLDPBD216361-250MG
tert-Butyl (1-(hydroxymethyl)cyclobutyl)carbamate 95%
tert-Butyl (1-(hydroxymethyl)cyclobutyl)carbamate

Storage condition: Sealed in dry, 2-8°C

Formule: C₁₀H₁₉NO₃
MW: 201,2628 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD12028429
CAS-nummer: 1142211-17-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR