3,5-Dimethyl-4-fluorobenzonitrile 98%

Leverancier: BLD PHARMATECH GMBH

Synoniemen: 4-Fluoro-3,5-dimethylbenzonitrile

BLDPBD217302-10GEA 191 EUR
BLDPBD217302-10G BLDPBD217302-250MG BLDPBD217302-5G BLDPBD217302-1G
3,5-Dimethyl-4-fluorobenzonitrile 98%
3,5-Dimethyl-4-fluorobenzonitrile

Storage condition: Sealed in dry, Room Temperature

Formule: C₉H₈FN
MW: 149,1700 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD09832248
CAS-nummer: 867367-02-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR