4-Phenyl-1H-pyrazole 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD218427-1GEA 299 EUR
BLDPBD218427-1G BLDPBD218427-250MG BLDPBD218427-5G BLDPBD218427-100MG
4-Phenyl-1H-pyrazole 95%
4-Phenyl-1H-pyrazole

Storage condition: Sealed in dry, Room Temperature

Formule: C₉H₈N₂
MW: 144,17 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00462204
CAS-nummer: 10199-68-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR