2-Chlorobenzylamine 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD219181-10GEA 9 EUR
BLDPBD219181-10G BLDPBD219181-5G BLDPBD219181-25G BLDPBD219181-1G
2-Chlorobenzylamine 98%
2-Chlorobenzylamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₇H₈ClN
MW: 141,6 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00008108
CAS-nummer: 89-97-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR