D-Alanine ethyl ester hydrochloride 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD21922-1GEA 0 EUR
BLDPBD21922-1G BLDPBD21922-100G BLDPBD21922-10G BLDPBD21922-500G BLDPBD21922-25G BLDPBD21922-5G BLDPBD21922-1000G
D-Alanine ethyl ester hydrochloride 98%
D-Alanine ethyl ester hydrochloride

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₅H₁₂ClNO₂
MW: 153,61 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00190725
CAS-nummer: 6331-09-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR