5-Bromopyrimidin-2-ol 98%

BLDPBD2196-5GEA 14 EUR
BLDPBD2196-5G BLDPBD2196-10G BLDPBD2196-100G BLDPBD2196-25G BLDPBD2196-500G
5-Bromopyrimidin-2-ol 98%
5-Bromopyrimidin-2-ol

Storage condition: Sealed in dry, Room Temperature

Formule: C₄H₃BrN₂O
MW: 174,9800 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00233952
CAS-nummer: 214290-49-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR