N,N'-Bis(3-methylphenyl)-N,N'-bis(phenyl)benzidine 97%

BLDPBD22028-25GEA 74 EUR
BLDPBD22028-25G BLDPBD22028-100G BLDPBD22028-1G BLDPBD22028-5G
N,N'-Bis(3-methylphenyl)-N,N'-bis(phenyl)benzidine 97%
N,N'-Bis(3-methylphenyl)-N,N'-bis(phenyl)benzidine

Storage condition: Sealed in dry, Room Temperature

Formule: C₃₈H₃₂N₂
MW: 516,6741 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00144965
CAS-nummer: 65181-78-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR