(3,4-Dichlorophenyl)acetic acid methyl ester 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD22120-1GEA 9 EUR
BLDPBD22120-1G BLDPBD22120-25G BLDPBD22120-5G BLDPBD22120-10G BLDPBD22120-100G
(3,4-Dichlorophenyl)acetic acid methyl ester 98%
(3,4-Dichlorophenyl)acetic acid methyl ester

Storage condition: Sealed in dry, Room Temperature

Formule: (C₆H₃Cl₂)CH₂COOCH₃
MW: 219,07 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00800680
CAS-nummer: 6725-44-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR