1-Boc-piperidine 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD22507-25GEA 42 EUR
BLDPBD22507-25G BLDPBD22507-5G BLDPBD22507-10G BLDPBD22507-1G
1-Boc-piperidine 98%
1-Boc-piperidine

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₀H₁₉NO₂
MW: 185,27 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD02093935
CAS-nummer: 75844-69-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR