Isopropyl-4-hydroxypiperidine-1-carboxylate 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD22780-10GEA 48 EUR
BLDPBD22780-10G BLDPBD22780-5G BLDPBD22780-1G
Isopropyl-4-hydroxypiperidine-1-carboxylate 98%
Isopropyl-4-hydroxypiperidine-1-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formule: C₉H₁₇NO₃
MW: 187,2400 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD08460168
CAS-nummer: 832715-51-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR