trans-4-Dimethylaminocrotonic acid hydrochloride 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD22836-1GEA 7 EUR
BLDPBD22836-1G BLDPBD22836-100G BLDPBD22836-10G BLDPBD22836-25G BLDPBD22836-500G BLDPBD22836-5G BLDPBD22836-250MG
trans-4-Dimethylaminocrotonic acid hydrochloride 97%
trans-4-Dimethylaminocrotonic acid hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formule: C₆H₁₂ClNO₂
MW: 165,62 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD03695466
CAS-nummer: 848133-35-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR