4-Methoxypiperidine 97%

BLDPBD228603-1GEA 11 EUR
BLDPBD228603-1G BLDPBD228603-25G BLDPBD228603-5G BLDPBD228603-100G BLDPBD228603-10G
4-Methoxypiperidine 97%
4-Methoxypiperidine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₆H₁₄ClNO
MW: 115,1700 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD04115010
CAS-nummer: 4045-24-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR