1-(3-Bromophenylsulphonyl)pyrrolidine 98%

BLDPBD229299-5GEA 106 EUR
BLDPBD229299-5G BLDPBD229299-25G BLDPBD229299-0.025 BLDPBD229299-1G
1-(3-Bromophenylsulphonyl)pyrrolidine 98%
1-(3-Bromophenylsulphonyl)pyrrolidine

Storage condition: Inert atmosphere, Room Temperature

Formule: C₁₀H₁₂BrNO₂S
MW: 290,1768 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD07363829
CAS-nummer: 214210-14-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR