2-Acetyl-3-fluoropyridine 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD22944-10GEA 166 EUR
BLDPBD22944-10G BLDPBD22944-25G BLDPBD22944-1G BLDPBD22944-250MG BLDPBD22944-5G
2-Acetyl-3-fluoropyridine 98%
2-Acetyl-3-fluoropyridine

Storage condition: Inert atmosphere, Room Temperature

Formule: C₇H₆FNO
MW: 139,1300 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD06659495
CAS-nummer: 87674-20-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR