4-Bromo-2-piperazinopyridine 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD229794-100MGEA 81 EUR
BLDPBD229794-100MG
4-Bromo-2-piperazinopyridine 97%
4-Bromo-2-piperazinopyridine
Formule: C₉H₁₂BrN₃
MW: 242,1157 g/mol
MDL-nummer: MFCD16618975
CAS-nummer: 1201643-59-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR