4-(4-Carboxypiperidine-1-carbonyl)phenylboronic acid 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD229974-5GEA 340 EUR
BLDPBD229974-5G BLDPBD229974-1G BLDPBD229974-250MG
4-(4-Carboxypiperidine-1-carbonyl)phenylboronic acid 98%
4-(4-Carboxypiperidine-1-carbonyl)phenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Formule: C₁₃H₁₆BNO₅
MW: 277,0800 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD07363784
CAS-nummer: 850593-02-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR