N-(4-Methoxybenzyl) 5-bromopicolinamide 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD230140-5GEA 640 EUR
BLDPBD230140-5G BLDPBD230140-250MG BLDPBD230140-1G
N-(4-Methoxybenzyl) 5-bromopicolinamide 98%
N-(4-Methoxybenzyl) 5-bromopicolinamide

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₁₄H₁₃BrN₂O₂
MW: 321,1692 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD09801060
CAS-nummer: 951885-02-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR