3-Methyl-1H-pyrazol-5(4H)-one 97%

BLDPBD2305-10GEA 6 EUR
BLDPBD2305-10G BLDPBD2305-100G BLDPBD2305-25G
3-Methyl-1H-pyrazol-5(4H)-one 97%
3-Methyl-1H-pyrazol-5(4H)-one

Storage condition: Sealed in dry, Room Temperature

Formule: C4H6N2O
MW: 98,11 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00020699
CAS-nummer: 108-26-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR