4-Methoxy-3-(piperidin-1-ylsulphonyl)phenylboronic acid 95%

BLDPBD230531-100MGEA 39 EUR
BLDPBD230531-100MG BLDPBD230531-1G BLDPBD230531-5G BLDPBD230531-250MG
4-Methoxy-3-(piperidin-1-ylsulphonyl)phenylboronic acid 95%
4-Methoxy-3-(piperidin-1-ylsulphonyl)phenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Formule: C₁₂H₁₈BNO₅S
MW: 299,1500 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD08235037
CAS-nummer: 874219-18-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR