3-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester 96%

Leverancier: BLD PHARMATECH GMBH

BLDPBD230794-100MGEA 10 EUR
BLDPBD230794-100MG BLDPBD230794-1G BLDPBD230794-5G BLDPBD230794-250MG
3-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester 96%
3-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Formule: C₁₆H₂₃BO₄
MW: 290,1624 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD08704682
CAS-nummer: 1198615-70-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR