3-Chloor-p-toluidine 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD23253-25GEA 8 EUR
BLDPBD23253-25G
3-Chloor-p-toluidine 98%
3-Chloor-p-toluidine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₇H₈ClN
MW: 141,6 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00007773
CAS-nummer: 95-74-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR