α,α,α-Trifluoro-m-cresol 98%

BLDPBD23308-10GEA 6 EUR
BLDPBD23308-10G BLDPBD23308-1000G BLDPBD23308-25G BLDPBD23308-500G BLDPBD23308-100G
α,α,α-Trifluoro-m-cresol 98%
α,α,α-Trifluoro-m-cresol

Storage condition: Inert atmosphere, Room Temperature

Formule: C₇H₅F₃O
MW: 162,11 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00002299
CAS-nummer: 98-17-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR