3-Isopropylmorpholine 95+%

BLDPBD233819-250MGEA 46 EUR
BLDPBD233819-250MG BLDPBD233819-100MG BLDPBD233819-1G
3-Isopropylmorpholine 95+%
3-Isopropylmorpholine

Storage condition: Sealed in dry, 2-8°C

Formule: C7H15NO
MW: 129,2 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD09054775
CAS-nummer: 927802-40-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR