1,3-Dimethyl-1H-indazol-5-amine 97%

BLDPBD234790-1GEA 97 EUR
BLDPBD234790-1G BLDPBD234790-250MG BLDPBD234790-5G
1,3-Dimethyl-1H-indazol-5-amine 97%
1,3-Dimethyl-1H-indazol-5-amine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formule: C₉H₁₁N₃
MW: 161,2 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD20705804
CAS-nummer: 5757-85-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR