1-Heptanesulphonic acid sodium salt monohydrate 98%

BLDPBD235726-25GEA 29 EUR
BLDPBD235726-25G BLDPBD235726-100G BLDPBD235726-5G
1-Heptanesulphonic acid sodium salt monohydrate 98%
1-Heptanesulphonic acid sodium salt monohydrate

Storage condition: Inert atmosphere, Room Temperature

Formule: C₇H₁₅NaO₃S·1H₂O
MW: 220,27 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00149550
CAS-nummer: 207300-90-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR