Methyl benzo[d]thiazole-7-carboxylate 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD236964-100MGEA 57 EUR
BLDPBD236964-100MG BLDPBD236964-5G BLDPBD236964-1G BLDPBD236964-250MG
Methyl benzo[d]thiazole-7-carboxylate 95%
Methyl benzo[d]thiazole-7-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formule: C₉H₇NO₂S
MW: 193,22 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD13190218
CAS-nummer: 1038509-28-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR