Thiomorpholine-1,1-dioxide hydrochloride 97%

BLDPBD2385-5GEA 18 EUR
BLDPBD2385-5G BLDPBD2385-10G BLDPBD2385-1G BLDPBD2385-100G BLDPBD2385-25G
Thiomorpholine-1,1-dioxide hydrochloride 97%
Thiomorpholine-1,1-dioxide hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formule: C₄H₁₀ClNO₂S
MW: 171,6457 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD06738995
CAS-nummer: 59801-62-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR